Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500315

Sigma-Aldrich

Anti-TRIAD3A antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 359.00

CHF 359.00


Estimated to ship on25 May 2025



Select a Size

Change View
100 μG
CHF 359.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 359.00


Estimated to ship on25 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 95 kDa

species reactivity

mouse, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNF216(54476)

Related Categories

General description

Ring finger protein 216 (RNF216) is an E3 ubiquitin ligase. The gene encoding this protein is localized on human chromosome 7p22.3.

Immunogen

TRIAD3A antibody was raised against a peptide corresponding to 15 amino acids near the amino-terminus of mouse TRIAD3A.

Biochem/physiol Actions

Ring finger protein 216 (RNF216) associates with toll-like receptors and aids in their degradation. It also aids in the degradation of receptor interacting protein-1 (RIP-1) kinase and negatively modulates tumor necrosis α receptor signaling. The protein acts as an inhibitor of autophagy in macrophages. Mutations in the RNF216 gene have been associated with Gordon Holmes syndrome.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500315.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ubiquitylation of an internalized killer cell Ig-like receptor by Triad3A disrupts sustained NF-?B signaling.
Miah SM
Journal of Immunology (2011)
Regulation of autophagy by E3 ubiquitin ligase RNF216 through BECN1 ubiquitination.
Xu C
Autophagy (2014)
RNF216 mutations as a novel cause of autosomal recessive Huntington-like disorder.
Santens P
Neurology (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service