Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500288

Sigma-Aldrich

Anti-CUEDC1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CUE Domain-Containing Protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 406.00

CHF 406.00


Estimated to ship on02 June 2025



Select a Size

Change View
100 μG
CHF 406.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 406.00


Estimated to ship on02 June 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CUEDC1(404093)

Immunogen

CUEDC1 antibody was raised against a 16 amino acid peptide near the carbocy terminus of human CUEDC1.

Biochem/physiol Actions

CUEDC1 (CUE domain containing 1) might be involved in protein trafficking and degradation. Changes in this gene are linked with early stage cervical cancer. It is highly methylated in the placental tissue of preeclampsia patients. CUEDC1 is suggested to control trophoblast invasion by participating in the TGF (transforming growth factor)-β/Smad (mothers against decapentaplegic homolog) signaling pathway. The gene is mapped to human chromosome 17q23.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500288.

Physical form

Supplied in PBS with 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y-H Yan et al.
European review for medical and pharmacological sciences, 17(22), 3083-3094 (2013-12-05)
Preeclampsia is a complication of pregnancy that severely threatens the health of the mother and infant, yet the mechanism of pathogenesis remains unclear. In this article, gene array technology was applied to identify the genes related to the pathogenesis of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service