Skip to Content
Merck
All Photos(2)

Key Documents

SAB2700021

Sigma-Aldrich

Anti-Capsid Protein antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Capsid, Dengue virus 2 Capsid protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 337.00

CHF 337.00


Estimated to ship on27 April 2025



Select a Size

Change View
100 μL
CHF 337.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 337.00


Estimated to ship on27 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

Dengue virus

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
western blot: 500-3000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Immunogen

Synthetic peptide containing a sequence corresponding to a region within amino acids 2 and 13 of Capsid Protein (Dengue virus 2 ) according to NP_739591

Application

Suggested starting dilutions are as follows: FACS: Assay-dependent dilution, ICC/IF: 1:1000-1:2000, WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 1% BSA, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service