Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500372

Sigma-Aldrich

Anti-EXOC7 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Exo70p, Anti-Exocyst complex component 7, Anti-HGNC:23214

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 386.00

CHF 386.00


Estimated to ship on23 May 2025



Select a Size

Change View
100 μG
CHF 386.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 386.00


Estimated to ship on23 May 2025


biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EXOC7(23265)

General description

EXOC7 (Exocyst complex component 7) is a major component of a multiprotein, octameric complex, exocyst. It is localized in the active site of active exocytosis.

Immunogen

Peptide with sequence C-DNIKNDPDKEYN from the internal region of the protein sequence according to NP_001013861.1; NP_056034.2.

Application

Anti-EXOC7 antibody produced in goat is suitable for immunohistochemistry, indirect ELISA, and western blot analysis.

Biochem/physiol Actions

EXOC7 (Exocyst complex component 7) is mainly involved in the exocytosis, a membrane trafficking process of intracellular protein elements such as hormones and neurotransmitters, membrane proteins and lipids to specific domains of the plasma membrane. Exocytosis plays a vital role in the cellular growth, development and cell polarity establishment. During exocytosis, it directly connects with the plasma membrane through its direct interaction with phosphatidylinositol 4,5-bisphosphate (PI(4,5)P(2)). It has also been reported that the interaction between EXOC7 and PI(4,5)P(2) is highly essential for the docking and fusion of post-Golgi secretory vesicles.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianglan Liu et al.
Molecular biology of the cell, 18(11), 4483-4492 (2007-09-01)
The exocyst is an evolutionarily conserved octameric protein complex that tethers post-Golgi secretory vesicles at the plasma membrane for exocytosis. To elucidate the mechanism of vesicle tethering, it is important to understand how the exocyst physically associates with the plasma
Shu-Chan Hsu et al.
International review of cytology, 233, 243-265 (2004-03-24)
Exocytosis is an essential membrane traffic event mediating the secretion of intracellular protein contents such as hormones and neurotransmitters as well as the incorporation of membrane proteins and lipids to specific domains of the plasma membrane. As a fundamental cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service