Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500304

Sigma-Aldrich

Anti-DDX5/P68 RNA helicase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-DEAD/H box polypeptide 5, Anti-G17P1, Anti-HLR1, Anti-HUMP68

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 386.00

CHF 386.00


Estimated to ship on14 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1808


Select a Size

Change View
100 μG
CHF 386.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 386.00


Estimated to ship on14 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1808

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DDX5(1655)

General description

DDX5 (DEAD, Asp-Glu-Ala-Asp, box helicase 5) is a multifunctional protein belonging to the DEAD box family of RNA helicases. It consists of twelve conserved motifs (including the signature D-E-A-D motif) forming a conserved ′helicase′ central domain, which plays an important role in the RNA metabolism.
DEAD-box helicase 5 (DDX5) is encoded by the gene mapped to human chromosome 17q23.3–q24.2.

Immunogen

Peptide with sequence C-PMIGYPMPTGYSQ from the C Terminus of the protein sequence according to NP_004387.1.

Application

Anti-DDX5/P68 RNA helicase antibody produced in goat is suitable for indirect immunofluorescence, indirect ELISA, and western blot applications.

Biochem/physiol Actions

DDX5 (DEAD, Asp-Glu-Ala-Asp, box helicase 5) is associated with the regulation of pre-mRNA splicing mechanism. It acts as a transcriptional coactivator to control alternative splicing during the formation of spliceosome. DDX5 provides stability to the RNA secondary structure during several activities such as in the translation initiation, nuclear and mitochondrial splicing. It facilitates the unwinding as well as detachment of protein part from RNA. The protein also participates in the estrogen- and androgen-signaling pathway.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-resolution analysis of chromosomal imbalances using the Affymetrix 10K SNP genotyping chip.
Alexander H, et al
Genomics, 85(3), 392-400 (2005)
Samaan Samaan et al.
Nucleic acids research, 42(4), 2197-2207 (2013-11-28)
Estrogen and androgen receptors (ER and AR) play key roles in breast and prostate cancers, respectively, where they regulate the transcription of large arrays of genes. The activities of ER and AR are controlled by large networks of protein kinases
Frances V Fuller-Pace
Biochimica et biophysica acta, 1829(8), 756-763 (2013-03-26)
Members of the DEAD box family of RNA helicases, which are characterised by the presence of twelve conserved motifs (including the signature D-E-A-D motif) within a structurally conserved 'helicase' core, are involved in all aspects of RNA metabolism. Apart from
Ulrike Rappe et al.
The Journal of biological chemistry, 289(18), 12421-12434 (2014-03-20)
The armadillo repeat protein ARVCF is a component of adherens junctions. Similar to related proteins, such as p120-catenin and β-catenin, with known signaling functions, localization studies indicate a cytoplasmic and a nuclear pool of ARVCF. We find that ARVCF interacts

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service