Skip to Content
Merck
All Photos(6)

Key Documents

SAB2108934

Sigma-Aldrich

Anti-G6PC3 (N-terminal) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

TSG-6, TSG6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 323.00

CHF 323.00


Estimated to ship on06 May 2025



Select a Size

Change View
100 μL
CHF 323.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

CHF 323.00


Estimated to ship on06 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39 kDa

species reactivity (predicted by homology)

rat, bovine, mouse, human, rabbit, zebrafish, canine, guinea pig, horse

concentration

0.5 mg/mL

technique(s)

western blot: 1 μg/mL

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... G6PC3(57396)

General description

This gene encodes the catalytic subunit of glucose-6-phosphatase (G6Pase). G6Pase is located in the endoplasmic reticulum (ER) and catalyzes the hydrolysis of glucose-6-phosphate to glucose and phosphate in the last step of the gluconeogenic and glycogenolytic pathways. Mutations in this gene result in autosomal recessive severe congenital neutropenia. Multiple transcript variants have been found for this gene, only one of which is expected to express a protein.

Immunogen

Synthetic peptide directed towards the N-terminal region of Human G6PC3

Sequence

Synthetic peptide located within the following region: ESGYYSQAPAQVHQFPSSCETGPGSPSGHCMITGAALWPIMTALSSQVAT

Physical form

Supplied at 0.5 mg/ml in phosphate-buffered saline, 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service