Skip to Content
Merck
All Photos(2)

Key Documents

SAB2101262

Sigma-Aldrich

Anti-KIF2A (ab2) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-HK2, Anti-KIF2, Anti-Kinesin heavy chain member 2A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 356.00

CHF 356.00


Estimated to ship on02 May 2025



Select a Size

Change View
100 μL
CHF 356.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

CHF 356.00


Estimated to ship on02 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

80 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KIF2A(3796)

Immunogen

Synthetic peptide directed towards the N terminal region of human KIF2A

Biochem/physiol Actions

KIF2A belongs to the kinesin-like protein family, MCAK/KIF2 subfamily. It contains 1 kinesin-motor domain. KIF2A plus end-directed microtubule-dependent motor required for normal brain development. It may regulate microtubule dynamics during axonal growth. KIF2A is implicated in formation of bipolar mitotic spindles. It has microtubule depolymerization activity. Hela cells lacking KIF2A show asymmetric or monopolar mitotic spindles. Osteosarcoma cells (U2OS) lacking KIF2A or KIF2B show disorganised or monopolar mitotic spindles.Kinesins, such as KIF2, are microtubule-associated motor proteins. For background information on kinesins, see MIM 148760.[supplied by OMIM].

Sequence

Synthetic peptide located within the following region: IEPSPETPPPPASSAKVNKIVKNRRTVASIKNDPPSRDNRVVGSARARPS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service