Skip to Content
Merck
All Photos(2)

Key Documents

SAB2100604

Sigma-Aldrich

Anti-DNAJB1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DnaJ (Hsp40) homolog, subfamily B, member 1, Anti-HSPF1, Anti-Hdj1, Anti-Hsp40, Anti-Sis1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

38 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DNAJB1(3337)

General description

DnaJ (Hsp40) homolog, subfamily B, member 1 (DNAJB1) belongs to the heat shock protein 40 (HSP40) protein family. It contains a highly conserved J domain. The DNAJB1 gene is mapped to human chromosome 19p13.12.

Immunogen

Synthetic peptide directed towards the N terminal region of human DNAJB1

Application

Anti-DNAJB1 antibody produced in rabbit has been used in western blotting (1:500).

Biochem/physiol Actions

DnaJ (Hsp40) homolog, subfamily B, member 1 (DNAJB1) interacts with heat shock protein 70 (HSP70) and can stimulate its ATPase activity. It stimulates the association between heat shock cognate 70 kDa protein (HSC70) and Hsc70-interacting protein (HIP). In association with HSP70, HSP40 favors adenosine triphosphate (ATP)-dependent protein refolding, transport, and interaction. It acts as a negative regulator melanoma differentiation-associated gene 5 (MDA5) based mitochondrial antiviral signaling protein pathway and mitogen-inducible gene 6 (MIG6). It also blocks p53 mediated apoptosis by destabilizing programmed cell death 5 (PDCD5). Hsp40 mediates viral ribonucleoproteins (vRNPs) import and may serve as a potential antiviral target.

Sequence

Synthetic peptide located within the following region: GSGPSGGSGGGANGTSFSYTFHGDPHAMFAEFFGGRNPFDTFFGQRNGEE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Ting Feng et al.
BMC developmental biology, 19(1), 9-9 (2019-04-27)
Coilia nasus oogenesis/spawning migration is a well-defined synchronous arrangement process. DnaJs are indispensable molecular chaperones for oogenesis process. However, how DnaJs involved the anadromous spawning migration mechanism is outstanding and plausible. In this regard, two DnaJs (Cn-DnaJa1 and Cn-DnaJb1) are
Soo-Yeon Park et al.
Biochimica et biophysica acta, 1853(10 Pt A), 2722-2730 (2015-08-05)
Mitogen-inducible gene 6 (MIG6) is a tumor suppressor implicated in the development of human cancers; however, the regulatory mechanisms of MIG6 remain unknown. Here, using a yeast two-hybrid screen, we identified DnaJ homolog subfamily B member I (DNAJB1) as a
Monika Vyas et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 33(4), 648-656 (2019-11-05)
Recently discovered DNAJB1-PRKACA oncogenic fusions have been considered diagnostic for fibrolamellar hepatocellular carcinoma. In this study, we describe six pancreatobiliary neoplasms with PRKACA fusions, five of which harbor the DNAJB1-PRKACA fusion. All neoplasms were subjected to a hybridization capture-based next-generation
Jyoti Batra et al.
Scientific reports, 6, 19063-19063 (2016-01-12)
A unique feature of influenza A virus (IAV) life cycle is replication of the viral genome in the host cell nucleus. The nuclear import of IAV genome is an indispensable step in establishing virus infection. IAV nucleoprotein (NP) is known
Ken Takashima et al.
Journal of innate immunity, 10(1), 44-55 (2017-10-27)
Melanoma differentiation-associated gene 5 (MDA5) is a pattern recognition receptor that recognizes cytoplasmic viral double-stranded RNA (dsRNA) and initiates rapid innate antiviral responses. MDA5 forms a filament-like multimer along the dsRNA leading to oligomerization, which in turn activates the adaptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service