Skip to Content
Merck
All Photos(2)

Key Documents

SAB2100215

Sigma-Aldrich

Anti-BCL11A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-B-cell CLL/lymphoma 11A (zinc finger protein), Anti-BCL11A-L, Anti-BCL11A-S, Anti-BCL11A-XL, Anti-CTIP1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 427.00

CHF 427.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1051


Select a Size

Change View
100 μL
CHF 427.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

CHF 427.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1051

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

91 kDa

species reactivity

horse, mouse, rat, human, dog, bovine, pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BCL11A(53335)

Immunogen

Synthetic peptide directed towards the N terminal region of human BCL11A

Biochem/physiol Actions

BCL11A Is a C2H2 type zinc-finger protein by its similarity to the mouse Bcl11a/Evi9 protein. The corresponding mouse gene is a common site of retroviral integration in myeloid leukemia, and may function as a leukemia disease gene, in part, through its interaction with BCL6. During hematopoietic cell differentiation, this gene is down-regulated. It is possibly involved in lymphoma pathogenesis since translocations associated with B-cell malignancies also deregulates its expression.This gene encodes a C2H2 type zinc-finger protein by its similarity to the mouse Bcl11a/Evi9 protein. The corresponding mouse gene is a common site of retroviral integration in myeloid leukemia, and may function as a leukemia disease gene, in part, through its interaction with BCL6. During hematopoietic cell differentiation, this gene is down-regulated. It is possibly involved in lymphoma pathogenesis since translocations associated with B-cell malignancies also deregulates its expression. Multiple transcript variants encoding several different isoforms have been found for this gene.

Sequence

Synthetic peptide located within the following region: MSRRKQGKPQHLSKREFSPEPLEAILTDDEPDHGPLGAPEGDHDLLTCGQ

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service