Skip to Content
Merck
All Photos(1)

Key Documents

SAB1411931

Sigma-Aldrich

ANTI-SORCS3 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

SORCS, SORCS3

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
CHF 356.00

CHF 356.00


Check Cart for Availability


Select a Size

Change View
50 μG
CHF 356.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 356.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 135.8 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SORCS3(22986)

General description

This gene encodes one family member of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins. The VPS10 domain name comes from the yeast carboxypeptidase Y sorting receptor Vps10 protein. Members of this gene family are large with many exons but the CDS lengths are usually less than 3700 nt. Very large introns typically separate the exons encoding the VPS10 domain; the remaining exons are separated by much smaller-sized introns. These genes are strongly expressed in the central nervous system. Two of the five family members (sortilin and sortilin-related receptor) are synthesized as preproproteins; it is not yet known if this encoded protein is also a preproprotein. (provided by RefSeq)

Immunogen

SORCS3 (NP_055793.1, 1 a.a. ~ 1222 a.a) full-length human protein.

Sequence
MEAARTERPAGRPGAPLVRTGLLLLSTWVLAGAEITWDATGGPGRPAAPASRPPALSPLSPRAVASQWPEELASARRAAVLGRRAGPELLPQQGGGRGGEMQVEAGGTSPAGERRGRGIPAPAKLGGARRSRRAQPPITQERGDAWATAPADGSRGSRPLAKGSREEVKAPRAGGSAAEDLRLPSTSFALTGDSAHNQAMVHWSGHNSSVILILTKLYDFNLGSVTESSLWRSTDYGTTYEKLNDKVGLKTVLSYLYVNPTNKRKIMLLSDPEMESSILISSDEGATYQKYRLTFYIQSLLFHPKQEDWVLAYSLDQKLYSSMDFGRRWQLMHERITPNRFYWSVAGLDKEADLVHMEVRTTDGYAHYLTCRIQECAETTRSGPFARSIDISSLVVQDEYIFIQVTTSGRASYYVSYRREAFAQIKLPKYSLPKDMHIISTDENQVFAAVQEWNQNDTYNLYISDTRGIYFTLAMENIKSSRGLMGNIIIELYEVAGIKGIFLANKKVDDQVKTYITYNKGRDWRLLQAPDVDLRGSPVHCLLPFCSLHLHLQLSENPYSSGRISSKETAPGLVVATGNIGPELSYTDIGVFISSDGGNTWRQIFDEEYNVWFLDWGGALVAMKHTPLPVRHLWVSFDEGHSWDKYGFTSVPLFVDGALVEAGMETHIMTVFGHFSLRSEWQLVKVDYKSIFSRHCTKEDYQTWHLLNQGEPCVMGERKIFKKRKPGAQCALGRDHSGSVVSEPCVCANWDFECDYGYERHGESQCVPAFWYNPASPSKDCSLGQSYLNSTGYRRIVSNNCTDGLREKYTAKAQMCPGKAPRGLHVVTTDGRLVAEQGHNATFIILMEEGDLQRTNIQLDFGDGIAVSYANFSPIEDGIKHVYKSAGIFQVTAYAENNLGSDTAVLFLHVVCPVEHVHLRVPFVAIRNKEVNISAVVWPSQLGTLTYFWWFGNSTKPLITLDSSISFTFLAEGTDTITVQVAAGNALIQDTKEIAVHEYFQSQLLSFSPNLDYHNPDIPEWRKDIGNVIKRALVKVTSVPEDQILIAVFPGLPTSAELFILPPKNLTERRKGNEGDLEQIVETLFNALNQNLVQFELKPGVQVIVYVTQLTLAPLVDSSAGHSSSAMLMLLSVVFVGLAVFLIYKFKRKIPWINIYAQVQHDKEQEMIGSVSQSENAPKITLSDFTEPEELLDKELDTRVIGGIATIANSESTKEIPNCTSV

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service