Skip to Content
Merck
All Photos(2)

Key Documents

SAB1409867

Sigma-Aldrich

Monoclonal Anti-RASSF5 antibody produced in mouse

clone 5C2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC10823, MGC17344, Maxp1, NORE1, NORE1A, NORE1B, RAPL, RASSF3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 403.00

CHF 403.00


Estimated to ship on30 May 2025



Select a Size

Change View
100 μG
CHF 403.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 403.00


Estimated to ship on30 May 2025


biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5C2, monoclonal

form

buffered aqueous solution

mol wt

antigen 37.51 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RASSF5(83593)

General description

This gene is a member of the Ras association domain family. It functions as a tumor suppressor, and is inactivated in a variety of cancers. The encoded protein localizes to centrosomes and microtubules, and associates with the GTP-activated forms of Ras, Rap1, and several other Ras-like small GTPases. The protein regulates lymphocyte adhesion and suppresses cell growth in response to activated Rap1 or Ras. Multiple transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

RASSF5 (NP_113625, 100 a.a. ~ 206 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
DVRSIFEQPQDPRVPAERGEGHCFAELVLPGGPGWCDLCGREVLRQALRCTNCKFTCHPECRSLIQLDCSQQEGLSRDRPSPESTLTVTFSQNVCKPVEETQRPPTL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service