Skip to Content
Merck
All Photos(4)

Key Documents

SAB1408653

Sigma-Aldrich

Anti-ISG15 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

G1P2, IFI15, UCRP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 502.00

CHF 502.00


Estimated to ship on29 May 2025



Select a Size

Change View
100 μG
CHF 502.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 502.00


Estimated to ship on29 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~17.9 kDa

species reactivity

mouse, human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ISG15(9636)

General description

ISG15 is a ubiquitin-like protein that becomes conjugated to many cellular proteins upon activation by interferon-alpha (IFNA; MIM 147660) and -beta (IFNB; MIM 147640) (Zhao et al., 2005 [PubMed 16009940]).[supplied by OMIM

Immunogen

ISG15 (ENSP00000368699, 1 a.a. ~ 165 a.a) full-length human protein.

Sequence
MGWDLTVKMLAGNEFQVSLSSSMSVSELKAQITQKIGVHAFQQRLAVHPSGVALQDRVPLASQGLGPGSTVLLVVDKCDEPLSILVRNNKGRSSTYEVRLTQTVAHLKQQVSGLEGVQDDLFWLTFEGKPLEDQLPLGEYGLKPLSTVFMNLRLRGGGTEPGGRS

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caleb D Swaim et al.
Cell reports, 31(11), 107772-107772 (2020-06-20)
ISG15 is a ubiquitin-like modifier that also functions extracellularly, signaling through the LFA-1 integrin to promote interferon (IFN)-γ release from natural killer (NK) and T cells. The signals that lead to the production of extracellular ISG15 and the relationship between its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service