Skip to Content
Merck
All Photos(2)

Key Documents

SAB1406878

Sigma-Aldrich

Anti-KIAA0101 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

L5, NS5ATP9, OEATC-1, OEATC1, PAF, p15(PAF)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~12.32 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KIAA0101(9768)

General description

KIAA0101, also known as, proliferating cell nuclear antigen (PCNA) clamp associated factor (PCLAF), is a proliferating cell nuclear antigen-associated factor. It is a 15kDa protein. It is highly expressed in thymus and colon. KIAA0101 gene is located on the human chromosome 15q22.31.

Immunogen

KIAA0101 (NP_055551.1, 1 a.a. ~ 111 a.a) full-length human protein.

Sequence
MVRTKADSVPGTYRKVVAARAPRKVLGSSTSATNSTSVSSRKAENKYAGGNPVCVRPTPKWQKGIGEFFRLSPKDSEKENQIPEEAGSSGLGKAKRKACPLQPDHTNDEKE

Biochem/physiol Actions

KIAA0101, also known as, proliferating cell nuclear antigen (PCNA) clamp associated factor (PCLAF), helps in regulating DNA repair, cell cycle progression and cell proliferation. Overexpression of KIAA0101 inhibits UV-induced cell death. It is also used as a prognostic marker for hepatocellular carcinoma(HCC). Overexpression of KIAA0101 acts as a marker for adrenocortical carcinoma (ACC) and human gastric cancer.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gene expression profiling of Japanese psoriatic skin reveals an increased activity in molecular stress and immune response signals
Kulsk JK, et al.
Journal of Molecular Medicine, 83(12), 964-975 (2005)
KIAA0101 is overexpressed, and promotes growth and invasion in adrenal cancer
Jain M, et al.
PLoS ONE, 6(11), e26866-e26866 (2011)
Overexpression of KIAA0101 predicts high stage, early tumor recurrence, and poor prognosis of hepatocellular carcinoma
Yuan RH, et al.
Clinical Cancer Research, 13(18), 5368-5376 (2007)
Kun Zhu et al.
Cancer science, 104(3), 353-359 (2012-12-18)
Gastric cancer (GC) is one of the most common malignant tumors with a high rate of recurrence, which results in surgery being unsuccessful. Therefore, it is important to find the reason for the surgery failing. The purpose of the present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service