Skip to Content
Merck
All Photos(2)

Key Documents

SAB1403967

Sigma-Aldrich

Monoclonal Anti-IL1A, (C-terminal) antibody produced in mouse

clone 1F3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

IL-1A, IL1, IL1-ALPHA, IL1F1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 403.00

CHF 403.00


Check Cart for Availability


Select a Size

Change View
100 μG
CHF 403.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 403.00


Check Cart for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F3, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL1A(3552)

General description

The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine is a pleiotropic cytokine involved in various immune responses, inflammatory processes, and hematopoiesis. This cytokine is produced by monocytes and macrophages as a proprotein, which is proteolytically processed and released in response to cell injury, and thus induces apoptosis. This gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. It has been suggested that the polymorphism of these genes is associated with rheumatoid arthritis and Alzheimer′s disease. (provided by RefSeq)

Immunogen

IL1A (AAH13142, 172 a.a. ~ 271 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
KSSKDDAKITVILRISKTQLYVTAQDEDQPVLLKEMPEIPKTITGSETNLLFFWETHGTKNYFTSVAHPNLFIATKQDYWVCLAGGPPSITDFQILENQA

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service