Skip to Content
Merck
All Photos(3)

Key Documents

SAB1402263

Sigma-Aldrich

Monoclonal Anti-MARK3 antibody produced in mouse

clone 1A10, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 474.00

CHF 474.00


Estimated to ship on29 April 2025



Select a Size

Change View
100 μG
CHF 474.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

CHF 474.00


Estimated to ship on29 April 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A10, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MARK3(4140)

Immunogen

MARK3 (NP_002367.4, 402 a.a. ~ 500 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
HHKVQRSVFSSQKQRRYSDHAGPAIPSVVAYPKRSQTSTADSDLKEDGISSRKSSGSAVGGKGIAPASPMLGNASNPNKADIPERKKSSTVPSSNTASG

Biochem/physiol Actions

MARK3 (microtubule affinity regulating kinase 3) is activated by phosphorylation and is responsible for the phosphorylation of other proteins. Changes in the expression of MARK3 are associated with carcinogenesis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Su-Mei Xiao et al.
The Journal of clinical endocrinology and metabolism, 98(11), E1827-E1834 (2013-07-31)
Fifty-six genomic loci recently were identified as associated with bone mineral density (BMD) in a large meta-analysis study of mainly European-descent subjects. Circulating factors related to calcium and phosphate metabolism, eg, serum levels of calcium, phosphate, vitamin D metabolites, PTH

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service