Skip to Content
Merck
All Photos(1)

Key Documents

SAB1401927

Sigma-Aldrich

Anti-WNT3A antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC119418, MGC119419, MGC119420

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 474.00

CHF 474.00


Check Cart for Availability


Select a Size

Change View
100 μG
CHF 474.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 474.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WNT3A(89780)

General description

The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It encodes a protein which shows 96% amino acid identity to mouse Wnt3A protein, and 84% to human WNT3 protein, another WNT gene product. This gene is clustered with WNT14 gene, another family member, in chromosome 1q42 region. (provided by RefSeq)

Immunogen

WNT3A (AAI03922.1, 1 a.a. ~ 385 a.a) full-length human protein.

Sequence
MAPLGYFLLLCSLKQALGSYPIWWSLAVGPQYSSLGSQPILCASIPGLVPKQLRFCRNYVEIMPSVAEGIKIGIQECQHQFRGRRWNCTTVHDSLAIFGPVLDKATRESAFVHAIASAGVAFAVTRSCAEGTAAICGCSSRHQGSPGKGWKWGGCSEDIEFGGMVSREFADARENRPDARSAMNRHNNEAGRQAIASHMHLKCKCHGLSGSCEVKTCWWSQPDFRAIGDFLKDKYDSASEMVVEKHRESRGWVETLRPRYTYFKVPTERDLVYYEASPNFCEPNPETGSFGTRDRTCNVSSHGIDGCDLLCCGRGHNARAERRREKCRCVFHWCCYVSCQECTRVYDVHTCKNPGSRAGNSAHQPPHPQPPVRFHPPLRRAGKVP

Biochem/physiol Actions

During the differentiation of human neuronal progenitor cells, WNT3A (Wnt family member 3A) activates calmodulin dependent kinase II and increases free calcium in the cytosol via Wnt/Ca2+ signaling pathway. WNT3A is an oncogene and is upregulated in many types of cancer. WNT3A serves as a prognostic marker in diagnosing hepatocellular carcinoma.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cross-talk of WNT and FGF signaling pathways at GSK3beta to regulate beta-catenin and SNAIL signaling cascades.
Katoh M and Katoh M.
Cancer Biology & Therapy, Sep;5(9), 1059-1064 (2006)
Non-canonical pathway induced by Wnt3a regulates ?-catenin via Pyk2 in differentiating human neural progenitor cells.
Narendra Talabattula VA
Biochemical and Biophysical Research Communications, 491(1), 40-46 (2017)
Abnormality of Wnt3a expression as novel specific biomarker for diagnosis and differentiation of hepatocellular carcinoma.
Pan L
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 37(4), 5561-5568 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service