Skip to Content
Merck
All Photos(4)

Key Documents

SAB1401497

Sigma-Aldrich

Monoclonal Anti-RABGAP1L antibody produced in mouse

clone 2D3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp686E1450, HHL, TBC1D18

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 474.00

CHF 474.00


Estimated to ship on27 April 2025



Select a Size

Change View
100 μG
CHF 474.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 474.00


Estimated to ship on27 April 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RABGAP1L(9910)

Immunogen

RABGAP1L (NP_055672.3, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MEVRASLQKVSGSSDSVATMNSEEFVLVPQYADDNSTKHEEKPQLKIVSNGDEQLEKAMEEILRDSEKRPSSLLVDCQSSSEISDHSFGDIPASQTNKPSLQLILDPSNT

Biochem/physiol Actions

RABGAP1L is known to be associated with systemic lupus erythematosus and hypertension.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deletion variants of RABGAP1L, 10q21.3, and C4 are associated with the risk of systemic lupus erythematosus in Korean women.
Kim JH
Arthritis and Rheumatism, 65(4), 1055-1063 (2013)
Assessment of a polymorphism of SDK1 with hypertension in Japanese Individuals.
Oguri M
American Journal of Hypertension, 23(1), 70-77 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service