Skip to Content
Merck
All Photos(2)

Key Documents

SAB1401427

Sigma-Aldrich

Anti-RECK antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

ST15, hRECK

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 356.00

CHF 356.00


Check Cart for Availability


Select a Size

Change View
100 μG
CHF 356.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

CHF 356.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RECK(8434)

General description

The protein encoded by this gene is a cysteine-rich, extracellular protein with protease inhibitor-like domains whose expression is suppressed strongly in many tumors and cells transformed by various kinds of oncogenes. In normal cells, this membrane-anchored glycoprotein may serve as a negative regulator for matrix metalloproteinase-9, a key enzyme involved in tumor invasion and metastasis. (provided by RefSeq)

Immunogen

RECK (NP_066934.1, 1 a.a. ~ 971 a.a) full-length human protein.

Sequence
MATVRASLRGALLLLLAVAGVAEVAGGLAPGSAGALCCNHSKDNQMCRDVCEQIFSSKSESRLKHLLQRAPDYCPETMVEIWNCMNSSLPGVFKKSDGWVGLGCCELAIALECRQACKQASSKNDISKVCRKEYENALFSCISRNEMGSVCCSYAGHHTNCREYCQAIFRTDSSPGPSQIKAVENYCASISPQLIHCVNNYTQSYPMRNPTDSLYCCDRAEDHACQNACKRILMSKKTEMEIVDGLIEGCKTQPLPQDPLWQCFLESSQSVHPGVTVHPPPSTGLDGAKLHCCSKANTSTCRELCTKLYSMSWGNTQSWQEFDRFCEYNPVEVSMLTCLADVREPCQLGCRNLTYCTNFNNRPTELFRSCNAQSDQGAMNDMKLWEKGSIKMPFINIPVLDIKKCQPEMWKAIACSLQIKPCHSKSRGSIICKSDCVEILKKCGDQNKFPEDHTAESICELLSPTDDLKNCIPLDTYLRPSTLGNIVEEVTHPCNPNPCPANELCEVNRKGCPSGDPCLPYFCVQGCKLGEASDFIVRQGTLIQVPSSAGEVGCYKICSCGQSGLLENCMEMHCIDLQKSCIVGGKRKSHGTSFSIDCNVCSCFAGNLVCSTRLCLSEHSSEDDRRTFTGLPCNCADQFVPVCGQNGRTYPSACIARCVGLQDHQFEFGSCMSKDPCNPNPCQKNQRCIPKPQVCLTTFDKFGCSQYECVPRQLACDQVQDPVCDTDHMEHNNLCTLYQRGKSLSYKGPCQPFCRATEPVCGHNGETYSSVCAAYSDRVAVDYYGDCQAVGVLSEHSSVAECASVKCPSLLAAGCKPIIPPGACCPLCAGMLRVLFDKEKLDTIAKVTNKKPITVLEILQKIRMHVSVPQCDVFGYFSIESEIVILIIPVDHYPKALQIEACNKEAEKIESLINSDSPTLASHVPLSALIISQVQVSSSVPSAGVRARPSCHSLLLPLSLGLALHLLWTYN

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service