Skip to Content
Merck
All Photos(3)

Key Documents

SAB1400494

Sigma-Aldrich

Anti-ANKRD1 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-ALRP, Anti-C-193, Anti-CARP, Anti-CVARP, Anti-MCARP, Anti-bA320F15.2

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
CHF 473.00

CHF 473.00


Check Cart for Availability


Select a Size

Change View
50 μG
CHF 473.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

CHF 473.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANKRD1(27063)

General description

Ankyrin repeat domain-containing protein 1 (ANKRD1) is a member of muscle ankyrin repeat protein. It comprises a nuclear localization signal, four repeats of an ankyrin motif, and a PEST-like sequence. The ANKRD1 gene is mapped to human chromosome 10q23.31. It is expressed in skeletal and cardiac muscles.
The protein encoded by this gene is localized to the nucleus of endothelial cells and is induced by IL-1 and TNF-alpha stimulation. Studies in rat cardiomyocytes suggest that this gene functions as a transcription factor. Interactions between this protein and the sarcomeric proteins myopalladin and titin suggest that it may also be involved in the myofibrillar stretch-sensor system. (provided by RefSeq)

Immunogen

ANKRD1 (NP_055206.2, 1 a.a. ~ 319 a.a) full-length human protein.

Sequence
MMVLKVEELVTGKKNGNGEAGEFLPEDFRDGEYEAAVTLEKQEDLKTLLAHPVTLGEQQWKSEKQREAELKKKKLEQRSKLENLEDLEIIIQLKKRKKYRKTKVPVVKEPEPEIITEPVDVPTFLKAALENKLPVVEKFLSDKNNPDVCDEYKRTALHRACLEGHLAIVEKLMEAGAQIEFRDMLESTAIHWASRGGNLDVLKLLLNKGAKISARDKLLSTALHVAVRTGHYECAEHLIACEADLNAKDREGDTPLHDAVRLNRYKMIRLLIMYGADLNIKNCAGKTPMDLVLHWQNGTKAIFDSLRENSYKTSRIATF

Biochem/physiol Actions

Ankyrin repeat domain-containing protein 1 (ANKRD1) acts as a transcriptional regulator for cardiomyogenesis. It interacts with calsequestrin-2, intermediate filament protein desmin, and with Y-box transcription factor 1 (YB-1). Elevated expression of ANKRD1 is implicated in idiopathic dilated cardiomyopathy. The hepatitis C virus (HCV) coordinates with ANKRD1 for its entry and propagation. It serves as a co-activator of the p53 tumor suppressor protein and may regulate apoptosis in human ovarian cancer cells.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thoa T Than et al.
Scientific reports, 6, 20819-20819 (2016-02-11)
Hepatitis C virus (HCV) is highly dependent on host proteins for its own propagation. By transcriptome sequencing (RNA-Seq) analysis, we identified 30 host genes that were significantly differentially expressed in cell culture-grown HCV (HCVcc)-infected cells. Of these candidate genes, we
Akiko Takahashi et al.
Scientific reports, 8(1), 14896-14896 (2018-10-07)
Overcoming acquired resistance to epidermal growth factor receptor tyrosine kinase inhibitors (EGFR-TKIs) is critical in combating EGFR-mutant non-small cell lung cancer (NSCLC). We tried to construct a novel therapeutic strategy to conquer the resistance to second-and third-generation EGFR-TKIs in EGFR-positive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service