Skip to Content
Merck
All Photos(1)

Documents

SAB1300542

Sigma-Aldrich

Anti-FRAT1 (center) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-FRAT1, Anti-Frat-1, Anti-Frequently rearranged in advanced T-cell lymphomas, Anti-Proto-oncogene FRAT1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FRAT1(10023)

Related Categories

General description

FRAT1 belongs to the GSK-3-binding protein family. It may function in tumor progression and in lymphomagenesis. It positively regulates the Wnt signaling pathway by stabilizing beta-catenin through the association with GSK-3. It may play a role in tumor progression and collaborate with PIM1 and MYC in lymphomagenesis.

Immunogen

FRAT1 (NP_005470, 104-138)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the Center region of human FRAT1.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service