Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300524

Sigma-Aldrich

Anti-PIASz1 (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-KIAA1886, Anti-PIAS-like protein Zimp7, Anti-ZIMP7, Anti-ZMIZ2, Anti-Zinc finger MIZ domain-containing protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 299.00

CHF 299.00


Check Cart for Availability


Select a Size

Change View
100 μG
CHF 299.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 299.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PIASZ1(83637)

General description

PIASz is a members of the PIAS (protein inhibitor of activated signal transducer and activator of transcription) family of proteins, negative regulators of the Janus family of tyrosine kinase (JAK)-signal transducer and activator of transcription pathway. PIAS proteins have been shown to act as transcriptional co-regulators of multiple signaling pathways in various cellular processes. PIASz increases ligand-dependent transcriptional activity of androgen receptor and other nuclear hormone receptors, possibly through alteration of chromatin structure by SWI/SNF-like BAF complexes

Immunogen

PIASZ1 (NP_113637, 344-380)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human PIASz1.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service