Skip to Content
Merck
All Photos(2)

Key Documents

SAB1300412

Sigma-Aldrich

Anti-HS2ST1 (N-term) antibody produced in rabbit

Ig fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-2-O-sulfotransferase, Anti-2OST, Anti-HS2ST, Anti-HS2ST1, Anti-Heparan sulfate 2-O-sulfotransferase 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 339.00

CHF 339.00


Estimated to ship on28 May 2025



Select a Size

Change View
100 μG
CHF 339.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 339.00


Estimated to ship on28 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HS2ST1(9653)

General description

Heparan sulfate biosynthetic enzymes are key components in generating a myriad of distinct heparan sulfate fine structures that carry out multiple biologic activities. Heparan sulfate 2-O-sulfotransferase is a member of the heparan sulfate biosynthetic enzyme family. This family member transfers sulfate to the 2 position of the iduronic acid residue of heparan sulfate. The disruption of the HS2ST1 gene resulted in no kidney formation in knockout embryonic mice, indicating that the absence of this enzyme may interfere with the signaling required for kidney formation.

Immunogen

HS2ST1 (CAI19004, 21-55)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human HS2ST1.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service