Skip to Content
Merck
All Photos(1)

Key Documents

S9067

Sigma-Aldrich

Monoclonal Anti-Syntaxin 6 antibody produced in mouse

~2 mg/mL, clone 3D10, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3D10, monoclonal

form

buffered aqueous solution

mol wt

antigen ~29 kDa

species reactivity

rat, human, mouse

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
microarray: suitable
western blot: 2 μg/mL using S1 fraction of rat brain.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STX6(10228)
mouse ... Stx6(58244)
rat ... Stx6(60562)

General description

Monoclonal Anti-Syntaxin 6 (mouse IgG1 isotype) is derived from the 3D10 hybridoma produced by the fusion of NS-1 mouse myeloma cells and splenocytes from BALB/c mice. The syntaxin 6 (STX6) gene codes for a 255 amino acid protein. The protein is expressed at high level in brain, lung and kidney. STX6 has two regions, such as H1 and H2 domains. The H2 domain confers trans-Golgi localization. It belongs to the syntaxin family. STX6 gene is mainly expressed in endosomes and trans-Golgi network and is located on human chromosome 1q25.3.

Specificity

Monoclonal Anti-Syntaxin 6 recognizes rat, mouse, and human Syntaxin 6 (approx. 29 kDa).

Immunogen

syntaxin 6-GST fusion protein.

Application

Monoclonal Anti-Syntaxin 6 antibody produced in mouse may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunocytochemistry
  • enzyme-linked immunosorbent assay (ELISA)

Biochem/physiol Actions

Syntaxin 6 (STX6) protein participates in vesicular trafficking between the trans-Golgi network and endosomes or lysosomes. It interacts with the mammalian homolog of vacuolar protein sorting-associated protein 45 (VPS45), a sec1-like protein implicated in Golgi for prevacuolar compartment trafficking. STX6 plays a key role in controlling the caveolae-dependent endocytosis. It is actively involved in the maturation of endosome and GM1 (a ganglioside with one sialic acid) ganglioside membrane delivery. STX6 also plays a key role in reinternalization of glucose transporter-4 (Glu4) and sub endosomal Glu4 sorting. Overexpression of STX6 gene is observed in breast, tongue, colon, pancreatic, liver, prostate, skin, lung, cervical and gastric cancers. Antibodies, which react specifically with the syntaxin 6 protein are considered as an essential tool for studying vesicle trafficking in the cell.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at 2-8 °C for up to one month. For prolonged storage, freeze in working aliquots at −20 °C. Repeated freezing and thawing is not recommended. Storage in frost-free freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntaxin 6, a regulator of the protein trafficking machinery and a target of the p53 family, is required for cell adhesion and survival
Zhang Y, et al.
The Journal of Biological Chemistry, 283(45), 30689-30698 (2008)
The STX6-VTI1B-VAMP3 complex facilitates xenophagy by regulating the fusion between recycling endosomes and autophagosomes
Nozawa T, et al.
Autophagy, 13(1), 57-69 (2017)
Regulation of integrin endocytic recycling and chemotactic cell migration by syntaxin 6 and VAMP3 interaction
Riggs KA, et al.
Journal of Cell Science, 125(16), 3827-3839 (2012)
Syntaxin 6 functions in trans-Golgi network vesicle trafficking.
Bock JB, et al.
Molecular Biology of the Cell, 8(7), 1261-1271 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service