S5633
L-Serine benzyl ester hydrochloride
≥99% (TLC)
Synonym(s):
L-Serine phenylmethyl ester hydrochloride (1:1)
About This Item
Recommended Products
product name
L-Serine benzyl ester hydrochloride,
Assay
≥99% (TLC)
form
powder
color
white
storage temp.
−20°C
SMILES string
Cl.N[C@@H](CO)C(=O)OCc1ccccc1
InChI
1S/C10H13NO3.ClH/c11-9(6-12)10(13)14-7-8-4-2-1-3-5-8;/h1-5,9,12H,6-7,11H2;1H/t9-;/m0./s1
InChI key
MGZWCDQAKCHOBX-FVGYRXGTSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service