Skip to Content
Merck
All Photos(1)

Key Documents

S5304

Sigma-Aldrich

S-Ceramic HyperD® F

50 μm mean particle size

Synonym(s):

HyperD® ceramic ion exchangers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100

Quality Level

mean particle size

50 μm

capacity

>0.15 meq/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Ceramic HyperD media are used in protein chromatography and affinity chromatography. Ceramic HyperD media have been used to develop a chromatographic refolding process that allows processing of fusion peptides at a concentration range 10- to 100-fold higher than that observed for common refolding systems.

Features and Benefits

HyperD media are highly porous, rigid ceramic beads filled with a functionalized hydrophilic gel. The rigid bead allows extremely high linear flow rates without bed compression. The hydrogel exchanges throughout its volume, not just on the surface. And the media do not change volume due to changes in pH or ionic strength.
The supports are stable in commonly used solvents and alkaline or acid environments. Supplied as aqueous suspensions in 1 M NaCl with 20% ethanol to inhibit bacterial growth.

Legal Information

HyperD is a registered trademark of Pall Corporation

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F

Flash Point(C)

35 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nihal Tugcu et al.
Journal of chromatography. A, 954(1-2), 127-135 (2002-06-13)
In this paper, the selectivity of a variety of cation-exchange stationary phases was investigated using a homologous series of displacer molecules based on pentaerythritol. These displacers were derived from pentaerythritol and contained either four trimethyl ammonium groups [pentaerythrityl-(trimethylammonium chloride)4, PE(TMA)4]
Timothy M Pabst et al.
Journal of chromatography. A, 1216(45), 7950-7956 (2009-10-10)
This work provides a broad survey of binding and elution behavior of proteins on strong cation exchangers. Four proteins comprising two monoclonal antibodies, lysozyme, and cytochrome c were used as models in the investigation. Seven chromatography resins with different base
Calvin C Walker et al.
Toxicological sciences : an official journal of the Society of Toxicology, 95(1), 74-81 (2006-08-19)
A small fish model and surface-enhanced laser desorption/ionization time-of-flight mass spectrometry were used to investigate plasma protein expression as a means to screen chemicals for estrogenic activity. Adult male sheepshead minnows (Cyprinodon variegatus) were placed into aquaria for seawater control
Elisabeth Schmoeger et al.
Journal of chromatography. A, 1216(48), 8460-8469 (2009-10-27)
Refolding of proteins must be performed under very dilute conditions to overcome the competing aggregation reaction, which has a high reaction order. Refolding on a chromatography column partially prevents formation of the intermediate form prone to aggregation. A chromatographic refolding
Impact of ionic strength on adsorption capacity of chromatographic particles employed in separation of monoclonal antibodies.
Wrzosek, K., et al.
Chemical Papers, 64(4), 461-468 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service