Skip to Content
Merck
All Photos(1)

Key Documents

S3884

Sigma-Aldrich

Sauvagine

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C202H346N56O63S
CAS Number:
Molecular Weight:
4599.31
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

Assay

≥97% (HPLC)

storage temp.

−20°C

InChI key

ILCVKEBXPLEGOY-ZLFMSJRASA-N

Amino Acid Sequence

Glp-Gly-Pro-Pro-Ile-Ser-Ile-Asp-Leu-Ser-Leu-Glu-Leu-Leu-Arg-Lys-Met-Ile-Glu-Ile-Glu-Lys-Gln-Glu-Lys-Glu-Lys-Gln-Gln-Ala-Ala-Asn-Asn-Arg-Leu-Leu-Leu-Asp-Thr-Ile-NH2

General description

Sauvagine is a polypeptide derived from the frog skin. It belongs to the CRH (corticotropin releasing factor) family. Sauvagine is made of 40 amino acids. It may also be found in the brain of mammals.

Biochem/physiol Actions

A peptide related to CRF and urotensin 1 that was first isolated from the skin of the frog Phyllomedusa sauvagei. Potent releaser of ACTH and β-endorphin from the anterior pituitary.
Sauvagine helps to liberate ACTH (adrenocorticotropic hormone) from the pituitary cells of rat. It has several important functions in the cardiovascular, endocrine and renal systems.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Renda et al.
Archives of histology and cytology, 52 Suppl, 317-323 (1989-01-01)
The authors present an overview of the main amphibian peptide families mainly derived from the skin and mostly discovered by Erspamer and his associates. The studies of the peptides do not only promote the understanding of their chemical, metabolical and
Eric Gutknecht et al.
Journal of neurochemistry, 104(4), 926-936 (2007-11-03)
Endogenous expression of the corticotropin-releasing factor type 2a receptor [CRF2(a)] but not CRF2(b) and CRF2(c) was observed in higher passage cultures of human Y79 retinoblastoma cells. Functional studies further demonstrated an increase in CRF2(a) mRNA and protein levels with higher
Javier Santos et al.
Psychoneuroendocrinology, 33(9), 1248-1256 (2008-08-12)
Previously, we showed that corticotropin-releasing factor (CRF) injected i.p. mimicked epithelial responses to stress, both stimulating ion secretion and enhancing permeability in the rat colon, and mast cells were involved. However, the ability of CRF-sensitive mucosal/submucosal loops to regulate intestinal
Takeshi Nakamura et al.
American journal of physiology. Heart and circulatory physiology, 296(2), H325-H332 (2008-12-09)
Urocortin 3 (Ucn3) is a new member of the corticotropin-releasing factor (CRF) peptide family and is considered to be a specific and endogenous ligand for CRF type 2 receptors (CRF2Rs). The presence of CRF(2)Rs has been reported in the nucleus
Graham C Boorse et al.
General and comparative endocrinology, 146(1), 9-18 (2006-01-18)
Peptides of the corticotropin-releasing factor (CRF) family are expressed throughout the central nervous system (CNS) and in peripheral tissues where they play diverse roles in physiology, behavior, and development. Current data supports the existence of four paralogous genes in vertebrates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service