Skip to Content
Merck
All Photos(1)

Key Documents

S2763

Sigma-Aldrich

Anti-Sheep IgG (whole molecule) antibody produced in donkey

affinity isolated antibody, lyophilized powder

Synonym(s):

Donkey Anti-Sheep IgG, Sheep IgG Detection

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CHF 233.00
5 X 1 MG
CHF 844.00

CHF 233.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
CHF 233.00
5 X 1 MG
CHF 844.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

CHF 233.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

donkey

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

technique(s)

quantitative precipitin assay: suitable

storage temp.

2-8°C

target post-translational modification

unmodified

Application

Anti-Sheep IgG (whole molecule) antibody produced in donkey was covalently coupled with epoxy beads and used as control in immunoprecipitation.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunoprecipitation (1 paper)

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.

Physical form

Lyophilized from 0.01 M sodium phosphate, 0.015 M sodium chloride, pH 7.2.

Reconstitution

Reconstitute with 0.135 M sodium chloride.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agnieszka Gambus et al.
The Journal of biological chemistry, 286(13), 11855-11864 (2011-02-02)
In late mitosis and G1, Mcm2-7 are assembled onto replication origins to license them for initiation in the upcoming S phase. After initiation, Mcm2-7 provide helicase activity to unwind DNA at the replication fork. Here we examine the structure of
Soumya Palliyil
Methods in molecular biology (Clifton, N.J.), 1673, 325-352 (2017-11-14)
A number of bacteria use a class of chemical compounds called acyl-homoserine lactones (AHLs) as quorum sensing (QS) signals to coordinate their behavior at the population level, including pathogens like Pseudomonas aeruginosa. Blocking QS using antibodies is an attractive strategy
S Hashira et al.
Pediatrics international : official journal of the Japan Pediatric Society, 42(4), 337-342 (2000-09-15)
Maternal immunoglobulin G (IgG), transferred across the placenta to the fetus during intrauterine life, is an important component of the neonatal immunological defence mechanisms against infection. There is controversy with respect to differences in placental transfer of the different IgG

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service