Skip to Content
Merck
All Photos(1)

Key Documents

S2125

Sigma-Aldrich

(−)Scopolamine N-oxide hydrobromide

Synonym(s):

Hyoscine N-oxide hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO5 · HBr
CAS Number:
Molecular Weight:
400.26
EC Number:
MDL number:
UNSPSC Code:
12352103

SMILES string

Br.C[N+]1([O-])C2C[C@@H](CC1C3OC23)OC(=O)[C@H](CO)c4ccccc4

InChI

1S/C17H21NO5.BrH/c1-18(21)13-7-11(8-14(18)16-15(13)23-16)22-17(20)12(9-19)10-5-3-2-4-6-10;/h2-6,11-16,19H,7-9H2,1H3;1H/t11?,12-,13-,14+,15-,16+,18?;/m1./s1

InChI key

MGNNYKWRWHQLCR-RDYQINQZSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Competitive antagonist

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service