S1505
Succinic semialdehyde solution
15% in H2O
Synonym(s):
4-Oxobutanoic acid, 4-Oxobutyric acid, Succinaldehydic acid, Succinic semialdehyde monomer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HCOCH2CH2COOH
CAS Number:
Molecular Weight:
102.09
Beilstein:
1745187
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
concentration
15% in H2O
storage temp.
−20°C
SMILES string
OC(=O)CCC=O
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Hongzhi Tang et al.
Applied and environmental microbiology, 74(5), 1567-1574 (2008-01-22)
Previous research suggested that Pseudomonas spp. may attack the pyrrolidine ring of nicotine in a way similar to mammalian metabolism, resulting in the formation of pseudooxynicotine, the direct precursor of a potent tobacco-specific lung carcinogen. In addition, the subsequent intermediates
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service