All Photos(1)
About This Item
Linear Formula:
C44H54O15N6
CAS Number:
Molecular Weight:
906.93
MDL number:
UNSPSC Code:
12171500
Assay
~90%
storage temp.
−20°C
SMILES string
N.N.N.N.COc1cc2cc(oc2cc1N3CCOCCOCCN(CCOCC3)c4cc5oc(cc5cc4OC)-c6ccc(cc6C(O)=O)C(O)=O)-c7ccc(cc7C(O)=O)C(O)=O
Application
Fluorescent indicator for intracellular sodium
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A Minta et al.
The Journal of biological chemistry, 264(32), 19449-19457 (1989-11-15)
Fluorescent indicators sensitive to cytosolic concentrations of free Na+ have been synthesized and characterized. They consist of a crown ether, 1,7-diaza-4,10,13-trioxacyclopentadecane, linked via its nitrogens to fluorophores bearing additional liganding centers. In the currently preferred dye, SBFI (short for sodium-binding
X Chen et al.
Biochemistry, 33(46), 13769-13774 (1994-11-22)
Although the predominance of plasmalogens in electrically-active membranes (e.g., sarcolemma) is well-known, identification of the molecular mechanisms through which the vinyl ether linkage facilitates electrophysiologic function has remained elusive. Herein we demonstrate that the kinetics of both carrier-mediated (i.e., valinomycin)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service