Key Documents
RFA453011-EP
Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
FA-45-30-11, aerosol tight, to hold, 30 x 1.5/2.0 mL tubes, lid
About This Item
UNSPSC Code:
41103909
Pricing and availability is not currently available.
feature
lid
to hold 30 x 1.5/2.0 mL tubes
aerosol tight
fixed angle rotor (microtubes)
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022637100 (NA)
Eppendorf® 5804726006 (EU)
parameter
20,800 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5804/5804R/5810/5810R
General description
A wide selections of rotors to fit the Eppendorf 5804 and 5810 centrifuges.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)