The Human RELT ELISA antibody pair and standard have been identified, but the kit is not fully developed. This ELISA requires a 4 - 7 week development phase, which begins after receiving an order for the kit. The standard curve range has not been determined at this time. After development, the kit will have passed quality control tests and will thereafter have a typical 1 - 2 day lead time for orders placed.
RAB1662
Human RELT ELISA
for serum, plasma and cell culture supernatants
Select a Size
CHF 791.00
Select a Size
About This Item
CHF 791.00
species reactivity
human
technique(s)
ELISA: suitable
input
sample type cell culture supernatant(s)
sample type plasma
sample type serum
assay range
inter-assay cv: <12%
intra-assay cv: <10%
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... RELT(84957)
Related Categories
General description
Application
Please refer to the attached Protocolfor details.
Other Notes
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Could you provide the protocol and range of the standard curve for the Human RELT ELISA Cat# RAB1662?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service