Skip to Content
Merck
All Photos(1)

Key Documents

RAB0808

Sigma-Aldrich

Mouse Il25 / Interleukin 25 ELISA Kit

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
CHF 502.00

CHF 502.00


Estimated to ship on29 May 2025



Select a Size

Change View
96 WELLS
CHF 502.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

CHF 502.00


Estimated to ship on29 May 2025


species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 50 pg/mL
standard curve range: 49.15-12000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il25(140806)

General description

The antibody pair provided in this kit recognizes mouse Interleukin 25.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arzu D Yalcin et al.
Current pharmaceutical design, 25(35), 3784-3795 (2019-10-01)
Multi-center, randomized-controlled trials and observational studies have demonstrated that, in severe asthmatic patients receiving omalizumab treatment, the frequency of exacerbations, the number of urgent adverse events, and the need for oral steroids tend to decrease. This study included a total

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service