Skip to Content
Merck
All Photos(2)

Key Documents

RAB0137

Sigma-Aldrich

Rat Thymus Chemokine-1 / CXCL7 ELISA Kit

for serum, plasma and cell culture supernatant

Synonym(s):

TCK-1

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
CHF 719.00

CHF 719.00


Estimated to ship on04 May 2025



Select a Size

Change View
1 KIT
CHF 719.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

CHF 719.00


Estimated to ship on04 May 2025


species reactivity

rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type plasma
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 80 pg/mL
standard curve range: 68.59-50000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

The Rat TCK-1 (Thymus Chemokine-1) ELISA (Enzyme- Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Rat TCK-1 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Rat Thymus Cytokine 1

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Pro-platelet basic protein (PPBP)/chemokine (C-X-C motif) ligand 7 (CXCL7) is a platelet-derived growth factor. It acts as an effective chemoattractant and neutrophil activator.[1] CXCL7 interacts with its receptor C-X-C motif chemokine receptor 1/2 to promote tumor angiogenesis and tumor cell migration.[1] Overexpression of the gene has been observed in peripheral blood cells of patients with renal cell carcinoma (RCC). Thus, it might be considered as a potent diagnostic marker for RCC.[1]

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression level of CXCL7 in peripheral blood cells is a potential biomarker for the diagnosis of renal cell carcinoma
Kinouchi T, et al.
Cancer Science, 108(12), 2495-2502 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service