Skip to Content
Merck
All Photos(2)

Key Documents

RAB0135

Sigma-Aldrich

Human NAP-2 / CXCL7 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
CHF 501.00

CHF 501.00


Check Cart for Availability


Select a Size

Change View
1 KIT
CHF 501.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

CHF 501.00


Check Cart for Availability

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type urine
sample type serum
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 8.5 pg/mL
standard curve range: 4.1-1000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PPBP(5473)

General description

Platelet-derived chemokine (C-X-C motif) ligand 7 (CXCL7), also known as neutrophil-activating protein-2 (NAP-2), is a member of a subset of seven chemokines. The protein is produced in platelets[1] and its structure contains an N-terminal glutamate-leucine-arginine (ELR) motif.[2] It acts as a ligand for the CXC chemokine receptor 2 (CXCR2).[3] NAP-2 facilitates the migration of bone marrow mesenchymal stem cells (MSCs) to the injury site.[1] In addition, it also aids in monocyte recruitment during atherogenesis.[3] The Human NAP-2 enzyme-linked immunosorbent assay (ELISA) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human NAP-2 in serum, plasma, cell culture supernatants, and urine.
The Human NAP-2 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human NAP-2 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human NAP2

Application

Human NAP-2 / CXCL7 ELISA Kit has been used in enzyme-linked immunosorbent assay to quantitatively measure NAP-2/CXCL7 in serum samples of chronic myelomonocytic leukemia (CMML) patients.[4]

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
For research use only. Not for use in diagnostic procedures.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Baltus et al.
Journal of leukocyte biology, 78(2), 435-441 (2005-05-19)
Platelet-derived chemokines, such as regulated on activation, normal T expressed and secreted (RANTES; CC chemokine ligand 5), platelet factor 4 [PF4; CXC chemokine ligand 4 (CXCL4)], and epithelial neutrophil-activating protein 78 (ENA-78; CXCL5), or precursors, such as beta-thromboglobulin, which can
Wei Jiang et al.
Experimental and therapeutic medicine, 19(4), 2449-2456 (2020-04-08)
Overuse and misuse of antibiotics leads to antibiotic resistance which has become a significant public health concern. Klebsiella pneumoniae is the most common pathogenic bacteria underlying nosocomial infections due to the expression of virulence factors and occurrence of antibiotic resistance.
Catarina R Almeida et al.
Stem cell reports, 6(4), 466-473 (2016-04-08)
Strategies for improved homing of mesenchymal stem cells (MSCs) to a place of injury are being sought and it has been shown that natural killer (NK) cells can stimulate MSC recruitment. Here, we studied the chemokines behind this recruitment. Assays
Aaron J Brown et al.
Frontiers in immunology, 8, 1248-1248 (2017-10-19)
Platelet-derived chemokine CXCL7 (also known as NAP-2) plays a crucial role in orchestrating neutrophil recruitment in response to vascular injury. CXCL7 exerts its function by activating the CXC chemokine receptor 2 (CXCR2) receptor and binding sulfated glycosaminoglycans (GAGs) that regulate
Qian Guo et al.
Oncology reports, 37(2), 1114-1122 (2016-12-14)
CXCL7 is an important chemoattractant cytokine, which signals through binding to its receptor CXCR2. Recent studies have demonstrated that the CXCL7/CXCR2 signaling plays a promoting role in several common malignancies, including lung, renal, colon, and breast cancer. However, the regulatory

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service