Skip to Content
Merck
All Photos(2)

Key Documents

RAB0086

Sigma-Aldrich

Mouse CD40 ELISA Kit

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
CHF 497.00

CHF 497.00


Check Cart for Availability


Select a Size

Change View
1 KIT
CHF 497.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

CHF 497.00


Check Cart for Availability

Quality Level

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 3 pg/mL
standard curve range: 2.06-1500 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Cd40(958)

General description

The Mouse CD40 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse CD40/TNFRSF5 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse CD40

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dennis Wolf et al.
Circulation, 129(23), 2414-2425 (2014-03-26)
Costimulatory cascades such as the CD40L-CD40 dyad enhance immune cell activation and inflammation during atherosclerosis. Here, we tested the hypothesis that CD40 directly modulates traits of the metabolic syndrome in diet-induced obesity in mice. To induce the metabolic syndrome, wild-type
Adem Aydın et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 32(1), 151-158 (2013-10-12)
In this study, analysis and comprehensive comparison of neurogenic differentiation capacity of human bursal tissue-derived-stem cells (hBT-SCs) was aimed with human bone marrow derived mesenchymal stem cells (hBM-MSCs). hBT-SCs was isolated from subacromial bursa tissue (n = 3) by collagen

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service