Skip to Content
Merck
All Photos(1)

Documents

R6891

Unwire full racks, polyketone

orange, Holds 40 x 20-mm tubes

Synonym(s):

moulded rack, test tube rack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122804
NACRES:
NB.35

material

polyketone

feature

Holds 40 x 20-mm tubes

packaging

pack of 1 ea

manufacturer/tradename

Nalgene® 5970-0120

L × W × H

250 mm × 102 mm × 83 mm

color

orange

Looking for similar products? Visit Product Comparison Guide

General description

Molded of durable polyketone. Unwire test tube racks have no coating that can chip or peel and permit corrosion. Excellent replacement for wire racks; withstands most lab chemicals. Excellent thermal properties and won′t get brittle in freezers. Grid design allows tubes to be tipped from side to side for clearly marking position. These racks will not float in water baths.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Unwire is a trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service