Skip to Content
Merck
All Photos(2)

Key Documents

R5405

Sigma-Aldrich

Anti-Rabex-5 (N-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-RAP1, Anti-Rab5 GDP/GTP exchange factor (RABGEF1), Anti-Rabaptin-5-associated exchange factor for Rab5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~58 kDa

species reactivity

mouse, rat, human

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 5-10 μg/mL using rat NRK cells fixed and permeabilized with 4% paraformaldehyde followed by 0.4% saponin
western blot: 2-5 μg/mL using whole extracts of mouse brain and human U87 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Anti-Rabex-5 (N-terminal) is produced in rabbit using as immunogen a synthetic peptide corresponding to a sequence at N-terminal of human Rabex-5, with an added cysteine, conjugated to KLH. Rabex-5 contains multiple functional domains, including a vacuolar protein sorting-associated (VPS9) domain that possesses the GDP/GTP exchange factors (GEF) activity, a ubiquitin-binding A20-like zinc finger (ZnF) domain, and a coiled-coil domain.

Application

Anti-Rabex-5 (N-terminal) antibody may be used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

Rabex-5 forms a tight complex with Rabaptin-5 that is essential for endocytic membrane fusion. The bound Rabaptin-5 to Rabex-5 increases the GEF activity of Rabex-5 and fastens the generation of the active GTP-bound form of membrane associated Rab5.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kavitha Balaji et al.
Traffic (Copenhagen, Denmark), 15(11), 1206-1218 (2014-08-02)
Listeria monocytogenes is a food-borne pathogenic bacterium that invades intestinal epithelial cells through a phagocytic pathway that relies on the activation of host cell RAB5 GTPases. Listeria monocytogenes must subsequently inhibit RAB5, however, in order to escape lysosome-mediated destruction. Relatively
Functional synergy between Rab5 effector Rabaptin-5 and exchange factor Rabex-5 when physically associated in a complex
Lippe R, et al.
Molecular Biology of the Cell, 12(7), 2219-2228 (2001)
The Rab5 guanine nucleotide exchange factor Rabex-5 binds ubiquitin (Ub) and functions as a Ub ligase through an atypical Ub-interacting motif and a zinc finger domain
Mattera R, et al.
Test, 281(10), 6874-6883 (2006)
The RAB5-GEF Function of RIN1 Regulates Multiple Steps During Listeria monocytogenes Infection.
Kavitha Balaji et al.
Traffic (Copenhagen, Denmark), 16(7), 796-796 (2015-06-09)
David Millrine et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(38), 10625-10630 (2016-09-08)
Immunomodulatory drugs (IMiDs) are a family of compounds derived from thalidomide. Binding of the IMiD molecule to the Lon protease Cereblon initiates the degradation of substrates via the ubiquitin proteasome pathway. Here, we show that Cereblon forms a complex with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service