Skip to Content
Merck
All Photos(1)

Key Documents

R4030

Sigma-Aldrich

Anti-hnRNP-E1 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HNRPX, Anti-PCBP1, Anti-hnRNP-X, Anti-poly(rC) binding protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~37 kDa

species reactivity

mouse, human, rat

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using K562 cell extract
western blot: 0.5-1 μg/mL using K562 cell extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCBP1(5093)
mouse ... Pcbp1(23983)
rat ... Pcbp1(500242)

General description

Heterogeneous nuclear ribonucleoprotein E1 (hnRNP E1)/poly(rC)-binding protein 1 (PCBP1) has five major isoforms, hnRNP-E1-E4 and hnRNP-E2-KL (a splice variant of hnRNP-E2). It is a member of the PCBP family or the triple K homology (KH) domain containing protein family. All isoforms of hnRNPE contain three repeats of the ribonucleoprotein KH domain, designated KH1, KH2 and KH3. hnRNP-E1 and hnRNP-E2 are predominantly nuclear with specific enrichment of hnRNP-E1 in nuclear speckles. Among the hnRNPE family, hnRNP-E1 and hnRNP-E2 share the highest amino acid sequence similarity. PCBP1 is ubiquitously expressed and is located on human chromosome 2p13.3.

Specificity

Anti-hnRNP-E1 (C-terminal) (also known as PCBP1/2 and αCP1) specifically recognizes human, mouse, and rat hnRNP-E1.

Application

Anti-hnRNP-E1 (C-terminal) antibody produced in rabbit may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Heterogeneous nuclear ribonucleoproteins (hnRNPs) are important in biological activities such as transcription, pre-mRNA processing, mRNA translation and turnover. hnRNPE proteins are also implicated in mRNA stabilization, translation enhancement and translation silencing. Heterogeneous nuclear ribonucleoprotein E1 (hnRNP E1)/poly(rC)-binding protein 1 (PCBP1) acts as a tumor-suppressive splicing factor. Upregulation of this gene can repress the tumorigenicity of ovarian cancer, prostate cancer, gastric cancer, colon cancer and thyroid carcinoma cells.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Common dysregulated genes in endometriosis and malignancies
Dentillo DB, et al.
Revista Brasileira de Ginecologia e obstetricia : Revista da Federac?o Brasileira das Sociedades de Ginecologia e Obstetricia, 38(5), 253-262 (2016)
PCBP1 inhibits the expression of oncogenic STAT3 isoform by targeting alternative splicing of STAT3 exon 23
Wang X, et al.
International Journal of Biological Sciences, 15(6), 1177-1177 (2019)
PCBP-1 regulates alternative splicing of the CD44 gene and inhibits invasion in human hepatoma cell line HepG2 cells
Zhang T, et al.
Molecular Cancer, 9(1), 1-10 (2010)
Margus Varjak et al.
Journal of virology, 87(18), 10295-10312 (2013-07-19)
Alphavirus replicase complexes are initially formed at the plasma membrane and are subsequently internalized by endocytosis. During the late stages of infection, viral replication organelles are represented by large cytopathic vacuoles, where replicase complexes bind to membranes of endolysosomal origin.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service