Skip to Content
Merck
All Photos(1)

Key Documents

R1281

Sigma-Aldrich

ROCK1 (17-535), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

P160ROCK, ROCK-I, ROKbeta

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CHF 486.00

CHF 486.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
CHF 486.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

CHF 486.00


Estimated to ship on21 May 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

49-67 nmol/min·mg

mol wt

~85 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ROCK1(6093)

Biochem/physiol Actions

ROCK1 is a ubiquitously expressed serine/threonine kinase that is a downstream target of the small GTPase RhoA. ROCK1 is involved in diverse cellular functions, including smooth muscle contraction, actin cytoskeleton organization, cell adhesion and motility, and gene expression.ROCK1 contributes to the development of cardiac fibrosis and induction of fibrogenic cytokines in cardiomyocytes in response to pathological stimuli. ROCK1 knockout mice exhibit reduced perivascular and interstitial fibrosis which is associated with reduced expression of a variety of extracellular matrix (ECM) proteins and fibrogenic cytokines.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying-Min Zhang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 20(7), 916-925 (2006-05-06)
Ventricular myocyte hypertrophy is an important compensatory growth response to pressure overload. However, pathophysiological cardiac hypertrophy is accompanied by reactive fibrosis and remodeling. The Rho kinase family, consisting of ROCK1 and ROCK2, has been implicated in cardiac hypertrophy and ventricular
Zhiyong Zhao et al.
Developmental biology, 275(1), 183-191 (2004-10-07)
Development of the endocardial cushions in the heart involves cell migration and cell differentiation, which is known as epithelial-mesenchymal transformation (EMT). These processes are regulated by cell signaling systems. Yet, the roles of intracellular GTPases and their effectors on these
Qing Zhou et al.
Hypertension (Dallas, Tex. : 1979), 69(6), 1070-1083 (2017-04-26)
Cardiac hypertrophy is characterized by increased myofibrillogenesis. Angiotensin II (Ang-II) is an essential mediator of the pressure overload-induced cardiac hypertrophy in part through RhoA/ROCK (small GTPase/Rho-associated coiled-coil containing protein kinase) pathway. FHOD3 (formin homology 2 domain containing 3), a cardiac-restricted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service