Skip to Content
Merck
All Photos(1)

Key Documents

PZ0354

Sigma-Aldrich

PF-04363467 hydrochloride

≥98% (HPLC)

Synonym(s):

(S)-N-[4-(4-Ethyl-2-morpholinyl)-2-methylphenyl]-4-(1-methylethyl)benzenesulfonamide hydrochloride, N-[4-[(2S)-4-Ethyl-2-morpholinyl]-2-methylphenyl]-4-(1-methylethyl)-benzenesulfonamide hydrochloride, PF-4363467 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 64.20
25 MG
CHF 315.00

CHF 64.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
CHF 64.20
25 MG
CHF 315.00

About This Item

Empirical Formula (Hill Notation):
C22H30N2O3S · HCl
CAS Number:
Molecular Weight:
439.01
UNSPSC Code:
51111800
NACRES:
NA.77

CHF 64.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 3 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

[H]Cl.CC1=CC([C@H]2CN(CC)CCO2)=CC=C1NS(C3=CC=C(C(C)C)C=C3)(=O)=O

InChI key

LDMGPGYFWNPBTL-VZYDHVRKSA-N

Biochem/physiol Actions

PF-4363467 hydrochloride is a Dopamine D3/D2 receptor antagonist. PF-4363467 has 100-fold selectivity for D3 with a Ki value of 3.1 nM for D3R compared to a Ki value of 692 nM for D2R and high selectivity for D3R compared to other biogenic amine receptors. PF-4363467 attenuated opioid drug-seeking behavior in a rat model of addiction without causing the extrapyramidal symptoms often associated with D2R antagonism.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service