Skip to Content
Merck
All Photos(1)

Key Documents

PZ0138

Sigma-Aldrich

SC-51089 hydrate

≥98% (HPLC)

Synonym(s):

8-Chloro-dibenz[b,f][1,4]oxazepine-10(11H)-carboxylic acid 2-[1-oxo-3-(4-pyridinyl)propyl]hydrazide monohydrochloride hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 117.00
25 MG
CHF 365.00

CHF 117.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
CHF 117.00
25 MG
CHF 365.00

About This Item

Empirical Formula (Hill Notation):
C22H19ClN4O3·HCl · xH2O
Molecular Weight:
459.33 (anhydrous basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

CHF 117.00


Estimated to ship on24 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥10 mg/mL

storage temp.

room temp

SMILES string

O.Cl.Clc1ccc2Oc3ccccc3CN(C(=O)NNC(=O)CCc4ccncc4)c2c1

InChI

1S/C22H19ClN4O3.ClH.H2O/c23-17-6-7-20-18(13-17)27(14-16-3-1-2-4-19(16)30-20)22(29)26-25-21(28)8-5-15-9-11-24-12-10-15;;/h1-4,6-7,9-13H,5,8,14H2,(H,25,28)(H,26,29);1H;1H2

InChI key

UEAWIGPAXPGSJC-UHFFFAOYSA-N

Biochem/physiol Actions

SC-51089 is a selective EP1 prostanoid receptor antagonist.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to prostanoid receptors for your research needs.

Discover Bioactive Small Molecules for Lipid Signaling Research

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service