Skip to Content
Merck
All Photos(1)

Key Documents

PP2386

Sigma-Aldrich

Yeast Tag Vector Set

plasmid vectors for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

conjugate

E-Tag
S-Tag
streptavidin conjugate

tag

10-His tagged
FLAG® tagged
GST tagged
Glu-Glu tagged
Hemagglutinin A (HA) tagged
Herpes Simplex Virus (HSV) tagged
T7 tagged
V5 tagged
c-Myc tagged
maltose-binding protein (MBP) tagged

form

buffered aqueous solution

bacteria selection

kanamycin

Origin of replication

2Micron
pUC (500 copies)

Peptide cleavage

no cleavage

Peptide tag location

N-terminal

Promoter

Promoter name: TEF1
Promoter activity: constitutive
Promoter type: yeast

shipped in

ambient

storage temp.

−20°C

General description

Molecular cloning often benefits from optimizing the vector used for expression.

This pack provides our entire range of 10 yeast functional tags, including epitope, affinity and solubility tags. Inserting your gene of interest into the MCS will place the tag downstream (at the C terminus of the protein), and the gene will be regulated by the strong constitutive yeast TEF1 promoter. This pack therefore provides a versatile resource for adding functional tags to several plasmids, but also enables you to compare different tags on the same protein, to identify which works best in your experimental system.This plasmid set has been designed to be compatible with a range of cloning techniques. The multiple cloning site contains a range of standard commonly used restriction sites for cloning. Using these sites genes can be inserted using standard cloning methods with DNA ligase. Other methods such as ligase independent cloning (LIC) Gibson Assembly InFusionHD or Seamless GeneArt can also be used and because all of our plasmids are based on the same backbone the same method can be used for cloning into all of our catalogue vectors.

Multiple cloning site notes: There are a few important sites within the MCS. These include the NcoI site the XbaI site and the BsgI and BseRI sites. The NcoI site contains a start codon that is immediately downstream of both a Kozak and Shine-Dalgarno ribosomal binding site. These allow for optimal positioning of genes when the start codon is placed in this location. If this is not required and you wish to use a downstream site for gene cloning you can remove the NcoI site by cleaving the plasmid with KpnI. The XbaI site contains a stop codon. This stop codon is positioned in a specific position in relation to the BsgI and BseRI sites that are immediately downstream. When either BseRI or BsgI cleave the plasmid they produce a TA overhang from the stop codon in the XbaI site that is compatible with all of our peptide tag plasmids cut with the same sites. BseRI and BsgI sites are non-palindromic and cleave a defined number of bases away from their binding site. Whenever we clone a gene into our multiple cloning site we always position the start and stop codon in the same positions in the MCS. If the start and ends of the genes are not compatible with NcoI and XbaI we extend the sequence to the nearest external sites but keep the start and stop codons locations consistent.

Transcription Termination: These plasmids contains three alternative transcription terminators for mammalian bacterial and bacteriophage (T7) expression. This means that only the promoter needs to be changed to alter the expression system you are using. We sell multiple promoters that can be used in each of these systems. The presence of each terminator does not reduce expression in the alternative systems.

Sequence

For Genebank sequence, FASTA sequence, Quick-reference Plasmid Map, and Full Plasmid Map, see the individual vector product page for links.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxfordgenetics.com.

Other Notes

Looking for more vector options to move your experiments forward faster? Consider a custom cloning vector designed and built by Oxford Genetics. Find out more at Oxford Genetics - Sigma's partner for cloning and expression vectors for molecular biology and synthetic biology applications.

Legal Information

These plasmids are sold free of reach-through rights and can be used to make commercial products. However the plasmids themselves (or derivatives) cannot be sold.
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
Oxford Genetics is a trademark of Oxford Genetics Ltd

Kit Components Also Available Separately

Product No.
Description
SDS

  • OGS3456PSF-TEF1-COOH-HIS6 - C-TERMINAL 6 HIS TAG YEAST PLASMID, plasmid vector for molecular cloningSDS

  • OGS3458PSF-TEF1-COOH-FLAG® - C-TERMINAL FLAG® TAG YEAST PLASMID, plasmid vector for molecular cloningSDS

  • OGS3459PSF-TEF1-COOH-CMYC - C-TERMINAL C-MYC TAG YEAST PLASMID, plasmid vector for molecular cloningSDS

  • PSF-TEF1-COOH-HA - C-TERMINAL HA TAG YEAST PLASMID, plasmid vector for molecular cloning

  • PSF-TEF1-COOH-GST - C-TERMINAL GST TAG YEAST PLASMID, plasmid vector for molecular cloning

  • PSF-TEF1-COOH-MBP - C-TERMINAL MBP TAG YEAST PLASMID, plasmid vector for molecular cloning

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana Romero et al.
Carcinogenesis, 37(1), 18-29 (2015-10-28)
Dickkopf-3 (Dkk-3) is a secreted protein whose expression is downregulated in many types of cancer. Endogenous Dkk-3 is required for formation of acini in 3D cultures of prostate epithelial cells, where it inhibits transforming growth factor (TGF)-β/Smad signaling. Here, we
Geoffrey M Lynn et al.
Nature biotechnology, 33(11), 1201-1210 (2015-10-27)
The efficacy of vaccine adjuvants such as Toll-like receptor agonists (TLRa) can be improved through formulation and delivery approaches. Here, we attached small molecule TLR-7/8a to polymer scaffolds (polymer-TLR-7/8a) and evaluated how different physicochemical properties of the TLR-7/8a and polymer
Alexander C Cerny et al.
PLoS genetics, 11(10), e1005578-e1005578 (2015-10-29)
Recycling of signaling proteins is a common phenomenon in diverse signaling pathways. In photoreceptors of Drosophila, light absorption by rhodopsin triggers a phospholipase Cβ-mediated opening of the ion channels transient receptor potential (TRP) and TRP-like (TRPL) and generates the visual
Jin-Gyoung Jung et al.
PLoS genetics, 10(10), e1004751-e1004751 (2014-10-31)
The Notch3 signaling pathway is thought to play a critical role in cancer development, as evidenced by the Notch3 amplification and rearrangement observed in human cancers. However, the molecular mechanism by which Notch3 signaling contributes to tumorigenesis is largely unknown.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service