Skip to Content
Merck
All Photos(3)

Documents

PLA0306

Sigma-Aldrich

Goat anti-USP7 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

HAUSP, Herpes virus-associated ubiquitin-specific protease, Herpesvirus-associated ubiquitin-specific protease, TEF1, deubiquitinating enzyme 7, ubiquitin specific protease 7 (herpes virus-associated), ubiquitin thioesterase 7, ubiquitin-specific-processing protease 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry: 1:1,000-1:5,000
immunoprecipitation (IP): 2-10 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_003461.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

goat ... USP7(7874)

Immunogen

The epitope recognized by PLA0306 maps to a region between residue 1 to 50 of human Ubiquitin Specific Peptidase 7 using the numbering given in entry NP_003461.1 (GeneID 7874).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Ubiquitin (Ub) is a highly conserved protein found ubiquitously in eukaryotic organisms. The conjugation of ubiquitin to proteins is an important means to regulate protein activity for many cellular processes by tagging them for degradation. Removal of Ub can rescue proteins from degradation. This is accomplished by the ubiquitin-specific processing protease (UBP) family of enzymes. Ubiquitin specific processing protease 7 (USP7) is a member of this family that has been shown to interact and stabilize p53.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaylor Boulay et al.
Life science alliance, 4(2) (2020-12-29)
Synovial sarcoma (SyS) is an aggressive mesenchymal malignancy invariably associated with the chromosomal translocation t(X:18; p11:q11), which results in the in-frame fusion of the BAF complex gene SS18 to one of three SSX genes. Fusion of SS18 to SSX generates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service