Skip to Content
Merck
All Photos(1)

Documents

PLA0253

Sigma-Aldrich

Rabbit anti-NPM1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

B23, NPM, member 1, nucleolar phosphoprotein B23, nucleolar protein NO38, nucleophosmin/nucleoplasmin family, numatrin, testicular tissue protein Li 128

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

immunoprecipitation (IP): 5-10 μg/mg

accession no.

NP_002511.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Gene Information

rabbit ... NPM1(4869)

Immunogen

The epitope recognized by PLA0253 maps to a region between residue 200 and 250 of human nucleophosmin using the numbering given in entry NP_002511.1 (GeneID 4869).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Nucleophosmin (NPM1) is involved in diverse cellular processes such as ribosome biogenesis, centrosome duplication, protein chaperoning, histone assembly, cell proliferation, and regulation of tumor suppressors TP53/p53 and ARF. It associates with nucleolar ribonucleoprotein structures and binds single-stranded nucleic acids. NPM1 binds ribosomes presumably to drive ribosome nuclear export. Chromosomal aberrations involving NPM1 have been identified in several types of leukemia [taken from the Universal Protein Resource (UniProt) www.uniprot.org/uniprot/P06748].

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Da Shi et al.
Scientific reports, 7, 39700-39700 (2017-01-04)
Porcine epidemic diarrhea virus (PEDV) replicates in the cytoplasm of infected cells, but its nucleocapsid (N) protein localizes specifically to the nucleolus. The mechanism of nuclear translocation, and whether N protein associates with particular nucleolar components, is unknown. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service