Inhibits DNA methyltransferase and modulates epigenetic regulation of gene expression. Na+ channel blocker and Class IA anti-arrhythmic.
Features and Benefits
This compound is a featured product for ADME Tox and Gene Regulation research. Discover more featured ADME Tox and Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Bristol-Myers Squibb. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.
Ventricular fibrillation (VF) is maintained by 2 mechanisms: first by reentry formation and second by spontaneous wave break or wave splitting. We hypothesized that spontaneous wave break results from a critical shortening of the action potential duration (APD) during VF
Chembiochem : a European journal of chemical biology, 13(1), 157-165 (2011-12-16)
DNA methyltransferases (DNMTs) are responsible for DNA methylation, an epigenetic modification involved in gene regulation. Families of conjugates of procainamide, an inhibitor of DNMT1, were conceived and produced by rapid synthetic pathways. Six compounds resulted in potent inhibitors of the
Chemical research in toxicology, 24(7), 1031-1039 (2011-06-16)
Aromatic amine drugs like aminoglutethimide (AG) and related congeners have been shown to produce phenyl radicals through metabolism by myeloperoxidase (MPO)/H(2)O(2), which has been proposed to play a role in drug-induced agranulocytosis. AG has also been shown to induce MPO
The Journal of chemical physics, 136(16), 164507-164507 (2012-05-09)
The pharmaceuticals, procaine hydrochloride and procainamide hydrochloride, are glass-forming as well as ionically conducting materials. We have made dielectric measurements at ambient and elevated pressures to characterize the dynamics of the ion conductivity relaxation in these pharmaceuticals, and calorimetric measurements
Procainamide, a type I antiarrhythmic agent, is used to treat a variety of atrial and ventricular dysrhythmias. It was reported that long-term therapy with procainamide may cause lupus erythematosus in 25-30% of patients. Interestingly, procainamide does not induce lupus erythematosus