Skip to Content
Merck
All Photos(1)

Key Documents

P8365

Sigma-Aldrich

Octakis (2,6-di-O-pentyl)-γ-cyclodextrin

>95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C128H240O40
Molecular Weight:
2419.25
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

>95%

form

crystals

technique(s)

electrophoresis: suitable

mp

<40 °C

storage temp.

2-8°C

SMILES string

CCCCCOC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](OCCCCC)[C@H](O[C@@H]3COCCCCC)O[C@H]4[C@H](O)[C@@H](OCCCCC)[C@H](O[C@@H]4COCCCCC)O[C@H]5[C@H](O)[C@@H](OCCCCC)[C@H](O[C@@H]5COCCCCC)O[C@H]6[C@H](O)[C@@H](OCCCCC)[C@H](O[C@@H]6COCCCCC)O[C@H]7[C@H](O)[C@@H](OCCCCC)[C@H](O[C@@H]7COCCCCC)O[C@H]8[C@H](O)[C@@H](OCCCCC)[C@H](O[C@@H]8COCCCCC)O[C@H]9[C@H](O)[C@@H](OCCCCC)[C@H](O[C@@H]9COCCCCC)O[C@H]1[C@H](O)[C@H]2OCCCCC

InChI

1S/C128H240O40/c1-17-33-49-65-137-81-89-105-97(129)113(145-73-57-41-25-9)121(153-89)162-106-90(82-138-66-50-34-18-2)155-123(115(98(106)130)147-75-59-43-27-11)164-108-92(84-140-68-52-36-20-4)157-125(117(100(108)132)149-77-61-45-29-13)166-110-94(86-142-70-54-38-22-6)159-127(119(102(110)134)151-79-63-47-31-15)168-112-96(88-144-72-56-40-24-8)160-128(120(104(112)136)152-80-64-48-32-16)167-111-95(87-143-71-55-39-23-7)158-126(118(103(111)135)150-78-62-46-30-14)165-109-93(85-141-69-53-37-21-5)156-124(116(101(109)133)148-76-60-44-28-12)163-107-91(83-139-67-51-35-19-3)154-122(161-105)114(99(107)131)146-74-58-42-26-10/h89-136H,17-88H2,1-16H3/t89-,90-,91-,92-,93-,94-,95-,96-,97+,98+,99+,100+,101+,102+,103+,104+,105-,106-,107-,108-,109-,110-,111-,112-,113-,114-,115-,116-,117-,118-,119-,120-,121-,122-,123-,124-,125-,126-,127-,128-/m1/s1

InChI key

FIGFUSWTQTYHTQ-YCWACCQZSA-N

Application

For capillary electrophoresis

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service