Skip to Content
Merck
All Photos(1)

Documents

P6660

Sigma-Aldrich

Phe-D-Met-Arg-Phe amide

≥97% (HPLC)

Synonym(s):

[D-Met2]-Molluscan cardioexcitatory neuropeptide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H42N8O4S
CAS Number:
Molecular Weight:
598.76
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

composition

Peptide content, ~65%

storage temp.

−20°C

SMILES string

CSCCC(NC(=O)C(N)Cc1ccccc1)C(=O)NC(CCCNC(N)=N)C(=O)NC(Cc2ccccc2)C(N)=O


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Mues et al.
Life sciences, 31(23), 2555-2561 (1982-12-06)
Phe-Met-Arg-Phe-NH2 (FMRFamide), injected at less than 1 mumol/kg intravenously in the anesthetized rat, produces sharp elevations of blood pressure and changes in respiration. The effects were dependent on the carboxyterminal Arg-Phe (RF) configuration and were stereospecific for these two amino

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service