Skip to Content
Merck
All Photos(1)

Key Documents

P5671

Sigma-Aldrich

N-(Phosphonomethyl)glycine

~95%

Synonym(s):

Glyphosate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2P(O)CH2NHCH2CO2H
CAS Number:
Molecular Weight:
169.07
Beilstein:
2045054
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

~95%

mp

230 °C (dec.) (lit.)

SMILES string

OC(=O)CNCP(O)(O)=O

InChI

1S/C3H8NO5P/c5-3(6)1-4-2-10(7,8)9/h4H,1-2H2,(H,5,6)(H2,7,8,9)

InChI key

XDDAORKBJWWYJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaofeng Gao et al.
Waste management & research : the journal of the International Solid Wastes and Public Cleansing Association, ISWA, 33(11), 1005-1014 (2015-09-30)
To evaluate the effects of different remediation methods on heavy metals contaminated recycled gravel, three immobilization agents (monopotassium phosphate, lime, nano-iron) and two mobilization agents (glyphosate, humic acid (HA)) were studied and compared. Results indicated that nano-iron powder was found
Antoine Mottier et al.
Marine pollution bulletin, 95(2), 665-677 (2014-12-03)
Glyphosate-based herbicides are extensively used and can be measured in aquatic ecosystems, including coastal waters. The effect of glyphosate on non-target organisms is an issue of worldwide concern. The aim of this study was to investigate the effects of subchronic
Emmanuel O Mogusu et al.
Analytical and bioanalytical chemistry, 407(18), 5249-5260 (2015-05-15)
To assess sources and degradation of the herbicide glyphosate [N-(phosphonomethyl) glycine] and its metabolite AMPA (aminomethylphosphonic acid), concentration measurements are often inconclusive and even (13)C/(12)C analysis alone may give limited information. To advance isotope ratio analysis of an additional element

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service