Skip to Content
Merck
All Photos(2)

Key Documents

P4659

Sigma-Aldrich

Poly(Lys, Tyr) hydrobromide

Lys:Tyr (4:1), mol wt 20,000-50,000

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.26

form

solid

feed ratio

Lys:Tyr (4:1)

mol wt

20,000-50,000

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaoqian Liu et al.
Biochemical and biophysical research communications, 339(3), 761-768 (2005-12-01)
Much attention has recently been paid to the study of positively charged polypeptides as a possible carrier for therapeutic protein or DNA delivery to cells. In this study, we have investigated the translocation of positively charged copoly(Lys/Tyr) (MW=72000, DP=385) across
Akira Shibata et al.
Biochimica et biophysica acta, 1616(2), 147-155 (2003-10-17)
Several membrane-transporting peptides (MTP) containing basic amino acid residues such as Lys and Arg that carry macromolecules such as DNA and proteins across cell plasma membranes by an unknown mechanism have been actively studied. On the basis of these results

Articles

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service